Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Silver trifluoromethanesulfonate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 8872209

 View more versions of this product

Catalog No. AA8872209



Specifications

Specifications

Silver trifluoromethanesulfonate
98%
MFCD00013226
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver;trifluoromethanesulfonate
76223
Powder
White
2923-28-6
CAgF3O3S
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
256.931
256.93
Odorless
10g

Chemical Identifiers

2923-28-6
256.931
QRUBYZBWAOOHSV-UHFFFAOYSA-M
76223
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
CAgF3O3S
MFCD00013226
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
silver;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

H314-H318-H400: Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 220-882-2

SDS Documents