missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver trifluoroacetate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1393306

 View more versions of this product

Catalog No. AA1393306



Specifications

Specifications

Silver trifluoroacetate
98%
MFCD00013199
KZJPVUDYAMEDRM-UHFFFAOYSA-M
silver;2,2,2-trifluoroacetate
76299
Powder
2966-50-9
C2AgF3O2
silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
C(=O)(C(F)(F)F)[O-].[Ag+]
220.883
220.88
5g

Chemical Identifiers

2966-50-9
220.883
KZJPVUDYAMEDRM-UHFFFAOYSA-M
76299
C(=O)(C(F)(F)F)[O-].[Ag+]
C2AgF3O2
MFCD00013199
silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
silver;2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

EINECSNumber : 221-004-0

Documents