missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver sulfide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4545806

 View more versions of this product

Catalog No. AA4545806



Specifications

Specifications

Silver sulfide
(metals basis)
MFCD00003406
14,8530
Pieces
7.317g/mL
6mm & down
825°C
21548-73-2
Ag2S
UN3077
247.8
99.999%
Odorless
Gray to Black
5g

Chemical Identifiers

21548-73-2
Ag2S
MFCD00003406
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents