Please login to your online account to display your discounted pricing

Silver Sulfate-Sulfuric Acid, For COD, LabChem™

$288.77 - $288.77

Chemical Identifiers

Chemical Name or Material Silver Sulfate-Sulfuric Acid
CAS 7664-93-9
CAS Max % 98.99
CAS 10294-26-5
CAS Max % 1.01
Name Note For COD
Linear Formula Ag2SO4
Formula Weight 311.79
UN Number UN1830
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC227601

 
lab chem
LC227601
500mL Each for $288.77
Description & Specifications

Specifications

Formula Weight 311.79
Decomposition Information Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Health Hazard 1 Danger
Packaging Amber Glass
Chemical Name or Material Silver Sulfate-Sulfuric Acid
Solubility Soluble in water
Linear Formula Ag2SO4
CAS Max % 1.01
Physical Form Liquid
Name Note For COD
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
CAS 7664-93-9
Quantity 500mL
CAS 10294-26-5
CAS Max % 98.99
Health Hazard 3 GHS P Statement
Do not breathe mist.
Wear eye protection, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
Color Clear, colorless to slight yellow
Storage Information Room Temperature
UN Number UN1830
Safety and Handling

Danger

GHS P Statement
Do not breathe mist.
Wear eye protection, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.