Please login to your online account to display your discounted pricing

Silver Sulfate, ACS Grade, ≥ 98.0%, LabChem™

$292.86 - $292.86

Chemical Identifiers

Chemical Name or Material Silver Sulfate
CAS 10294-26-5
CAS Max % 100
Purity Grade ACS Grade
Linear Formula Ag2SO4
Formula Weight 311.79
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC227558

 
lab chem
LC227558
25g Each for $292.86
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 311.79
Chemical Name or Material Silver Sulfate
EINECS Number 233-653-7
Identification Passes Test
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Crystalline
Solubility Soluble in water
Specific Gravity 5.45g/cm3
Storage Information Room Temperature
Linear Formula Ag2SO4
CAS 10294-26-5
Decomposition Information Sulfur compounds
Color White
Melting Point 652°C
Quantity 25g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Density 5.45g/cm3
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.