missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver sulfate, ACS, 98% , Alfa Aesar™

Manufacturer:  Alfa Aesar 1141714

 View more versions of this product

Catalog No. AA1141714



Specifications

Specifications

Silver sulfate
10294-26-5
Ag2O4S
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
311.792
311.8
5.45g/mL
For AAS analysis
98%
MFCD00003407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
disilver;sulfate
159865
Powder
25g

Chemical Identifiers

10294-26-5
311.792
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
159865
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
Ag2O4S
MFCD00003407
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
disilver;sulfate
Safety and Handling

Safety and Handling

EINECSNumber : 233-653-7

Documents