missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver sulfate, ACS reagent, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 194070100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silver sulfate | |
10g | |
Ag2SO4 | |
15,8668 | |
Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh,nitric acid and sulfuric acid,insoluble in alcohol | |
[O-]S(=O)(=O)[O-].[Ag+].[Ag+] | |
311.79 | |
311.79 | |
Glass bottle |
10294-26-5 | |
Ag2O4S | |
01,1015; 03,254; 04,435 | |
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
YPNVIBVEFVRZPJ-UHFFFAOYSA-L | |
disilver;sulfate | |
159865 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 233-653-7
RUO â Research Use Only