missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLAG22M
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Silver Standard | |
AgNO3 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver;nitrate | |
24470 | |
Triple-leached LDPE bottle | |
30 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
7761-88-8 | |
MFCD00003414 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
2% HNO3 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
CHEBI:32130 |
AgNO3 | |
MFCD00003414 | |
24470 | |
[Ag+].[O-][N+]([O-])=O |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient