Please login to your online account to display your discounted pricing

Silver Standard for ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 500mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLAG23X

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLAG2-3X

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Health Hazard 1 Causes severe skin burns and eye damage May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction Suspected of causing cancer.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Purity Grade Assurance Grade
Viscosity [NULL]
CAS 7732-18-5
Chemical Name or Material Silver Standard
CAS 7697-37-2
CAS 7440-22-4

Specifications

Chemical Name or Material Silver Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes severe skin burns and eye damage May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction Suspected of causing cancer.

Ambient

SDS & Safety Documents