Please login to your online account to display your discounted pricing

Silver Standard for ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$293.00 - $509.37

Chemical Identifiers

Chemical Name or Material Silver Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLAG2-3X

  SDS
spex certiprep llc
PLAG23X
Triple-leached LDPE bottle 500mL Each for $509.37

PLAG2-3Y

  SDS
spex certiprep llc
PLAG23Y
Triple-leached LDPE bottle 125mL Each for $293.00
Description & Specifications

Specifications

Quantity 500mL
Health Hazard 1 Causes severe skin burns and eye damage May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction Suspected of causing cancer.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Purity Grade Assurance Grade
Viscosity [NULL]
CAS 7732-18-5
Chemical Name or Material Silver Standard
CAS 7697-37-2
CAS 7440-22-4

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag10,000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes severe skin burns and eye damage May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction Suspected of causing cancer.

Ambient