missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver shot, 99.999% (metals basis), Alfa Aesar™, Premion™

Manufacturer:  Alfa Aesar 1415318

 View more versions of this product

Catalog No. AA1415318



Specifications

Specifications

Silver
7440-22-4
99.999% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
107.868
CHEBI:9141
Silver
50g
Silver-colored shot
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
Characteristic
silver
23954
Shot, 2-3mm (0.08-0.12 in.)

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

SDS Documents