missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver shot, 1-7mm (0.04-0.1in), Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
For vapor deposition
$237.01 - $3744.68
Chemical Identifiers
CAS | 7440-22-4 |
---|---|
Molecular Formula | Ag |
Molecular Weight (g/mol) | 107.87 |
MDL Number | MFCD00003397 |
InChI Key | BQCADISMDOOEFD-UHFFFAOYSA-N |
Synonym | argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber |
PubChem CID | 23954 |
ChEBI | CHEBI:9141 |
IUPAC Name | silver |
SMILES | [Ag] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1135709
|
Thermo Scientific Chemicals
01135709 |
10 g |
Each for $237.01
|
|
|||||
AA1135718
|
Thermo Scientific Chemicals
01135718 |
50 g |
Each for $906.81
|
|
|||||
AA1135730
|
Thermo Scientific Chemicals
01135730 |
250 g |
Each for $3,744.68
|
|
|||||
Description
For vapor deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Specifications
7440-22-4 | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
Premion™ | |
Shot | |
(metals basis) |
Ag | |
10 g | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
99.999% | |
Characteristic | |
Silver shot, 1 to 3mm (0.04 to 0.1 in.) |
Safety and Handling
EINECSNumber : 231-131-3
RTECSNumber : VW3500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only