missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver rod, 7mm (0.275 in.) dia., Premion™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 11473DH

 View more versions of this product

Catalog No. AA11473DH



Specifications

Specifications

Silver rod
25mm
7440-22-4
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Characteristic
23954
Rod
Premion™
≈4.04g/cm
7mm (0.275 in.) dia.
(metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
99.999%

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents