Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver rod, 6.35mm (0.25 in.) dia. x 150mm (5.9 in.) long, hard, Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Silver rod
CAS 7440-22-4
Name Note 6.35mm (0.25 in.) dia. x 150mm (5.9 in.) long, Hard
Purity Grade Premion™
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13304KG

  SDS
alfa aesar™
13304KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 piece
Purity Grade Premion™
TSCA TSCA
Chemical Name or Material Silver rod
Name Note 6.35mm (0.25 in.) dia. x 150mm (5.9 in.) long, Hard
Assay Percent Notes (metals basis)
Assay 99.99%
CAS 7440-22-4
MDL Number MFCD00003397
Weight ≈3.32g/cm
Odor Characteristic
Physical Form Rod