Please login to your online account to display your discounted pricing

Silver, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Ag) Solute: Silver Nitrate Solvent: Distilled Water

Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLSS457500

 View more versions of this product

Catalog No. SS457-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Density 1.0g/cm³
Vapor Pressure 14mmHg at 20°C
CAS 7732-18-5
Chemical Name or Material Silver, Reference Standard Solution
ChemAlert Storage Symbol White
CAS 7697-37-2
CAS 7761-88-8
MDL Number MFCD00003397
Packaging Glass Bottle
Traceability to NIST 3151 SILVER REF STD
Concentration 990 to 1010ppm
Name Note 1000ppm ±1%

Specifications

Chemical Name or Material Silver, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7761-88-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00003397
Accessories
 

SDS & Safety Documents