Please login to your online account to display your discounted pricing

Silver, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Ag) Solute: Silver Nitrate Solvent: Distilled Water

$76.94 - $124.00

Chemical Identifiers

Chemical Name or Material Silver, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7761-88-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS457-100

  SDS
fisher scientific company
SS457100
Glass Bottle 100mL Each for $76.94
Case of 6 Each for $461.64

SS457-500

  SDS
fisher scientific company
FLSS457500
Glass Bottle 500mL Each for $124.00
Case of 6 Each for $744.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
MDL Number MFCD00003397
CAS 7697-37-2
Packaging Glass Bottle
CAS 7732-18-5
Chemical Name or Material Silver, Reference Standard Solution
Name Note 1000ppm ±1%
Concentration 990 to 1010ppm
CAS 7761-88-8
ChemAlert Storage Symbol White
Traceability to NIST 3151 SILVER REF STD

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling