Please login to your online account to display your discounted pricing

Silver powder, spherical, APS 0.6-2 micron, 99.9% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silver powder
CAS 7440-22-4
Name Note spherical, APS 0.6 to 2μm
MDL Number MFCD00003397
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4159809

 
alfa aesar
4159809
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Chemical Name or Material Silver powder
TSCA TSCA
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00003397
Surface Area 0.4 to 1.5m2/g
Name Note spherical, APS 0.6 to 2μm
CAS 7440-22-4
UN Number UN3077
Odor Characteristic
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.