Learn More
Silver powder, spherical, APS 0.6-2 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Form: S.A. 0.4-1.5m2/g
Supplier: Thermo Scientific Chemicals 04159818
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver | |
7440-22-4 | |
10.5g/cm3 | |
Ag | |
50 g | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
Characteristic |
Silver | |
Silver-colored powder | |
99.9% (Metals basis) | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
Spherical Powder |
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Safety and Handling
Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only