missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Powder, 4-7 micron, 99.9% (Metals basis), Alfa Aesar™

Form: S.A. 0.1-0.4m2/g

Manufacturer:  Alfa Aesar 1140230

 View more versions of this product

Catalog No. AA1140230



Specifications

Specifications

Silver
7440-22-4
99.9% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver
250g
Silver-colored powder
Ag
algaedyn
[Ag]
Characteristic
23954
Powder

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-131-3

Documents