missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver powder, -22 mesh, Premion™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1075622

 View more versions of this product

Catalog No. AA1075622



Specifications

Specifications

Silver powder
(metals basis)
MFCD00003397
algaedyn
[Ag]
107.868
CHEBI:9141
99.999%
−22 Mesh
100g
7440-22-4
Ag
UN3077
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Powder
Premion™
Characteristic

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Documents