missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Powder, 0.7-1.3 micron, 99.9% (Metals basis), Alfa Aesar™

Form: S.A. 0.8-1.2m2/g

Manufacturer:  Alfa Aesar 1445009

 View more versions of this product

Catalog No. AA1445009



Specifications

Specifications

Silver
7440-22-4
10.5g/cm3
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
Characteristic
silver
23954
Powder
10g
Silver-colored powder
99.9% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
107.868
CHEBI:9141
Silver

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.

H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-131-3

SDS Documents