Please login to your online account to display your discounted pricing

Silver plating solution, metal content ≈28.7g/L, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silver plating solution
Name Note Metal content ≈28.7g/L
Physical Form Liquid
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44067AK

  SDS
alfa aesar
44067AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44067K2

  SDS
alfa aesar
44067K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250mL
Color Colorless
DOT Information DOT Class: 6.1, Packaging Group: II
Chemical Name or Material Silver plating solution
Name Note Metal content ≈28.7g/L
Physical Form Liquid
Appearance Colourless liquid
MDL Number MFCD00003397
Health Hazard 1 H300-H310-H330-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Safety and Handling

H300-H310-H330-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.