missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13849AP

 View more versions of this product

Catalog No. AA13849AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Silver
7440-22-4
Ag
Nitric acid: 5%; Silver: 0.1%; Water: 94.9%
[Ag]
107.868
CHEBI:9141
23 hPa (17mm Hg) at 20°C
500mL
plasma standard solution
Colourless liquid
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Liquid
Colorless

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
CHEBI:9141
[Ag]
Ag
MFCD00003397
23954
silver
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

Documents