missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver phosphate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1141514

 View more versions of this product

Catalog No. AA1141514



Specifications

Specifications

Silver phosphate
99%
MFCD00014148
silberphosphat
[O-]P(=O)([O-])[O-].[Ag+].[Ag+].[Ag+]
418.574
418.58
6.37g/cm3 at 20°C
Gray to Black
7784-09-0
Ag3O4P
Yellow powder
FJOLTQXXWSRAIX-UHFFFAOYSA-K
trisilver;phosphate
159913
Powder
Odorless
25g

Chemical Identifiers

7784-09-0
418.574
FJOLTQXXWSRAIX-UHFFFAOYSA-K
159913
[O-]P(=O)([O-])[O-].[Ag+].[Ag+].[Ag+]
Ag3O4P
MFCD00014148
silberphosphat
trisilver;phosphate
Safety and Handling

Safety and Handling

H318-H400-H410: Causes serious eye damage. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 232-049-0

Documents