missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver perchlorate monohydrate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1162409

 View more versions of this product

Catalog No. AA1162409



Specifications

Specifications

Silver perchlorate monohydrate
(metals basis)
MFCD00149128
14,8523
IXFNJWPLHOWEPT-UHFFFAOYSA-M
silver;perchlorate;hydrate
15959344
Crystalline
2.800g/mL
Light Sensitive & Hygroscopic
485°C (decomposition)
14242-05-8
AgClH2O5
UN1481
acmc-1brvv
O.[O-]Cl(=O)(=O)=O.[Ag+]
225.329
225.34 (207.32 Anhydrous)
99.9%
Odorless
White
10g

Chemical Identifiers

14242-05-8
225.329
IXFNJWPLHOWEPT-UHFFFAOYSA-M
15959344
O.[O-]Cl(=O)(=O)=O.[Ag+]
AgClH2O5
MFCD00149128
acmc-1brvv
silver;perchlorate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P280-P273-P303+P361+P353-P305+P351+P338-P310
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents