Please login to your online account to display your discounted pricing

Silver perchlorate hydrate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC210211000

 View more versions of this product

Catalog No. AC210211000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (on dry substance)
Physical Form Crystals or Crystalline Powder
Formula Weight 207.32
Quantity 100g
Color White
Merck Index 10, 8358
Chemical Name or Material Silver perchlorate hydrate
Iron (Fe) 20ppm max.
Assay Percent Range 99%
Linear Formula AgClO4·xH2O
Chlorine (Cl) 10ppm max.
Molecular Formula AgClO4·xH2O
CAS 14242-05-8
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Insoluble in Water 0.005% max.
Sulfate 10ppm max.
MDL Number MFCD00149128
Water 10% max. (K.F.)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Silver perchlorate hydrate
Assay ≥98.5% (on dry substance)
Assay Percent Range 99%
Molecular Formula AgClO4·xH2O
Linear Formula AgClO4·xH2O
Formula Weight 207.32
MDL Number MFCD00149128
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

SDS & Safety Documents