missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver perchlorate hydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 210210050

 View more versions of this product

Catalog No. AC210210050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Silver perchlorate hydrate
14242-05-8
5 g
AgClO4
MFCD00149128
silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
YDHABVNRCBNRNZ-UHFFFAOYSA-M
silver;perchlorate;hydrate
15959344
99%
White
Crystalline Powder and/or Crystals
98.5% min. (on dry substance) (Argentometry)
AgClO4·xH2O
10, 8358
Solubility in water: 557g/100mL. Other solubilities: soluble in many organic solvents
[Ag+].[O-][Cl](=O)(=O)=O
207.31
207.32
Glass bottle

Chemical Identifiers

14242-05-8
207.31
YDHABVNRCBNRNZ-UHFFFAOYSA-M
15959344
AgClO4
MFCD00149128
silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
[Ag+].[O-][Cl](=O)(=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only