Please login to your online account to display your discounted pricing

Silver perchlorate, anhydrous, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silver perchlorate
CAS 7783-93-9
Name Note Anhydrous
Molecular Formula AgClO4
Formula Weight 207.32
MDL Number MFCD00003400
UN Number UN1481
Merck Index 14,8523
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4220914

  SDS
alfa aesar
4220914
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.32
Quantity 25g
Chemical Name or Material Silver perchlorate
TSCA TSCA
Odor Odorless
Name Note Anhydrous
Physical Form Crystalline
Sensitivity Light Sensitive and Hygroscopic
Molecular Formula AgClO4
MDL Number MFCD00003400
CAS 7783-93-9
UN Number UN1481
Merck Index 14,8523
Color White
Melting Point 485°C (Dec.)
Solubility Information Soluble in water and many organic solvents (benzene, toluene, pyridine)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P221-P280-P273-P303+P361+P353-P305+P351+P338-P310
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Density 2.806
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P221-P280-P273-P303+P361+P353-P305+P351+P338-P310
Take any precaution to avoid mixing with combustibles.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.