Learn More
Silver perchlorate, anhydrous, Thermo Scientific Chemicals
Light Sensitive & Hygroscopic
Supplier: Thermo Scientific Chemicals 04220914
Description
Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver perchlorate | |
Anhydrous | |
White | |
25 g | |
MFCD00003400 | |
14,8523 | |
Soluble in water and many organic solvents (benzene,toluene,pyridine)Soluble in water,benzene and toluene. | |
[O-]Cl(=O)(=O)=O.[Ag+] | |
207.314 | |
207.32 | |
Hygroscopic; Light sensitive |
485°C (decomposition) | |
7783-93-9 | |
Crystalline | |
AgClO4 | |
UN1481 | |
silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous | |
YDHABVNRCBNRNZ-UHFFFAOYSA-M | |
silver;perchlorate | |
24562 | |
Odorless | |
2.806 |
Chemical Identifiers
7783-93-9 | |
207.314 | |
YDHABVNRCBNRNZ-UHFFFAOYSA-M | |
24562 | |
[O-]Cl(=O)(=O)=O.[Ag+] |
AgClO4 | |
MFCD00003400 | |
silver perchlorate, unii-sv4x1u113o, silver 1+ ion perchlorate, perchloric acid, silver 1+ salt, silver 1+ salt, ag.clo4, perchloric acid, silver 1+ salt 1:1, silver 1+ perchlorate ion, silver i perchloratehydrate, silver i perchlorate, anhydrous | |
silver;perchlorate |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 232-035-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only