Please login to your online account to display your discounted pricing

Silver oxide, 99%+, EMD Millipore™


Manufacturer: emd millipore corporation  1192080100

 View more versions of this product

Catalog No. M1192080100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 231.74
DOT Information DOT Class: 5.1 (8), Packaging Group: II
Health Hazard 1 H272: May intensify fire; oxidiser.
H314: Causes severe skin burns and eye damage.
EUH044: Risk of explosion if heated under confinement.
Physical Form Powder
Appearance Dark brown powder
Molecular Formula Ag2O (Hill)
Odor Odorless
Chemical Name or Material Silver oxide
Assay Percent Range 99%+
UN Number UN 3085
EINECS Number 243-957-1
Packaging Plastic bottle
CAS 20667-12-3

Specifications

Chemical Name or Material Silver oxide
CAS 20667-12-3
Molecular Formula Ag2O (Hill)
Formula Weight 231.74
UN Number UN 3085
Accessories

H272: May intensify fire; oxidiser.
H314: Causes severe skin burns and eye damage.
EUH044: Risk of explosion if heated under confinement.