Please login to your online account to display your discounted pricing

Silver Nitrate Standard Solution, 1mL = 1mg Chloride, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$240.48 - $861.78

Chemical Identifiers

Chemical Name or Material Silver Nitrate Standard Solution
CAS 7761-88-8
Name Note 1mL = 1mg Chloride
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula AgNO3
Formula Weight 169.87
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003927

 
avantor performance
565202
Narrow Mouth Amber Glass Bottle 1L Each for $240.48

02003928

 
avantor performance
565203
Narrow Mouth Amber Glass Bottle 4L Each for $861.78
Description & Specifications

Specifications

Formula Weight 169.87
Chemical Name or Material Silver Nitrate Standard Solution
Molecular Formula AgNO3
CAS 7761-88-8
Quantity 1L
Specific Gravity 1L = 1.00kg
Name Note 1mL = 1mg Chloride
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle