Learn More
Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Supplier: Thermo Scientific Chemicals 04308714
Description
Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver nitrate | |
| 7761-88-8 | |
| Crystalline | |
| (metals basis) | |
| MFCD00003414 | |
| 14,8518 | |
| Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether. | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 | |
| 99.995% | |
| Ag 63% min | |
| Light sensitive |
| 212°C | |
| White | |
| 25 g | |
| AgNO3 | |
| UN1479 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 | |
| 169.87 | |
| Premion™ | |
| Odorless | |
| 4.352 g/mL |
Chemical Identifiers
| 7761-88-8 | |
| 169.87 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| 24470 | |
| silver(1+) nitrate |
| AgNO3 | |
| MFCD00003414 | |
| silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
| CHEBI:32130 | |
| [Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H314-H290
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
May be corrosive to metals.
P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H290-H314-H335-H373
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 231-853-9
RTECSNumber : VW4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only