missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate-Nitric Acid, For Sulfate, Certified, LabChem™

$205.06 - $205.06

Chemical Identifiers

CAS 7732-18-5,7761-88-8,7697-37-2
CAS Max % 98.2,1.7,0.1
Linear Formula AgNO3
Formula Weight 169.87
Chemical Name or Material Silver Nitrate-Nitric Acid, For Sulfate
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC227501 Lab Chem
LC227501
500mL Each for $205.06
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7761-88-8,7697-37-2
AgNO3
Silver Nitrate-Nitric Acid, For Sulfate
98.2,1.7,0.1
169.87
Certified Grade

Specifications

7732-18-5,7761-88-8,7697-37-2
Liquid
169.87
Passes Test
Soluble in water
Passes Test
Nitrogen oxides; Silver
500mL
Certified Grade
98.2,1.7,0.1
AgNO3
1g/mL
Amber Glass
1g/mL
Room Temperature
Clear, colorless
Silver Nitrate-Nitric Acid, For Sulfate

Safety and Handling

GHS H Statement
Causes skin irritation and serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning