Please login to your online account to display your discounted pricing

Silver Nitrate, For Cyanide, (1mL = 1mg CN-), Certified, 0.0192N ±0.0002N (0.0192M), LabChem™

$64.17 - $96.89

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7732-18-5
CAS Max % 99.67
CAS 7761-88-8
CAS Max % 0.33
Name Note For Cyanide, (1mL = 1mg CN-)
Purity Grade Certified Grade
Linear Formula AgNO3
Formula Weight 169.87
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC226301

 
lab chem
LC226301
500mL Each for $64.17

LC226302

 
lab chem
LC226302
1L Each for $96.89
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 169.87
Chemical Name or Material Silver Nitrate
Traceability to NIST Traceable to NIST
Decomposition Information Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Cyanide, (1mL = 1mg CN-)
Physical Form Liquid
Concentration 0.0192N ±0.0002N (0.0192M)
Linear Formula AgNO3
Density 1g/mL
CAS 7732-18-5
CAS Max % 99.67
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 7761-88-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Amber Glass
CAS Max % 0.33
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.