missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, For Chloride, (1mL = 0.5mg Cl-), Certified, 0.0141N ±0.0002N (0.0141M), LabChem™

Manufacturer:  LabChem LC226001

 View more versions of this product

Catalog No. LC226001



Specifications

Specifications

Silver Nitrate
7761-88-8,7732-18-5
AgNO3
0.0141N ±0.0002N (0.0141M)
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Liquid
Passes Test
Soluble in water
Room Temperature
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide
500mL
For Chloride, (1mL = 0.5mg Cl-)
99.76,0.24
AgNO3
1g/mL
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
Certified
Amber Glass
1g/mL
Traceable to NIST
Colorless

Chemical Identifiers

7761-88-8
169.872
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
AgNO3
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver;nitrate
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Documents
Promotions

Promotions