Please login to your online account to display your discounted pricing

Silver Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$2,163.13 - $9,309.20

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7761-88-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula AgNO3
Formula Weight 169.87
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003929

 
avantor performance
342604
Wide Mouth Amber Glass Bottle 125g Case of 4 Each for $6,764.12

02003930

 
avantor performance
342600
Wide Mouth Amber Glass Bottle 30g Case of 4 Each for $2,163.13

02003931

 
avantor performance
342601
Wide Mouth Amber Glass Bottle 500g Each for $9,309.20
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 169.87
Chemical Name or Material Silver Nitrate
CAS 7761-88-8
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.35kg
Molecular Formula AgNO3
Quantity 125g