missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate (Cryst./Molecular Biology Grade), Fisher BioReagents

For silver staining

$290.40 - $290.40

Chemical Identifiers

CAS 7761-88-8
Physical Form Solid
Assay Percent Range ≥99.7 %
Molecular Formula AgNO3
Formula Weight 169.87
Synonym Nitric acid silver(I) salt
Chemical Name or Material Silver Nitrate
Assay ≥99.7%
Grade Molecular Biology Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BP2546100 SDS Fisher Scientific Company
BP2546100
Glass Bottle 100g Each for $290.40
Case of 6 Each for $1,742.40
Specifications

Description

Silver Nitrate is used in biochemistry to stain agarose and acrylamide gels. In histology, silver nitrate is used for silver staining to demonstrate reticular fibers, proteins and nucleic acids.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

7761-88-8
≥99.7 %
169.87
Silver Nitrate
Molecular Biology Grade
Solid
AgNO3
Nitric acid silver(I) salt
≥99.7%

Specifications

169.87
212°C
Solid
5.4 to 6.4
2ppm max.
≥99.7 %
Nitric acid silver(I) salt
7761-88-8
0.002% max.
5ppm max.
0.01% max.
Silver Nitrate
Molecular Biology Grade
444°C
100g
White
15, 8657
Pass Test
2ppm max.
0.001% max.
AgNO3
Glass Bottle
Pass Test
Pass Test
≥99.7%

Safety and Handling

DOTInformation : DOT Class 5.1, : Oxidizer

Recommended Storage : RT