missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, Certified, 10.0% (w/v) ±0.3%, LabChem™

Manufacturer:  LabChem LC225507

 View more versions of this product

Catalog No. LC225507



Specifications

Specifications

Silver Nitrate
90,10
AgNO3
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
10.0% ± 0.3% (w/v)
1.4g/mL
Amber Glass
1.4g/mL
Nitrogen oxides; Thermal decomposition generates: Corrosive vapors
125mL
7761-88-8,7732-18-5
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Liquid
Certified
Passes Test
Soluble in water
Room Temperature
Colorless

Chemical Identifiers

7761-88-8
169.872
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
AgNO3
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver;nitrate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe mist, spray, vapors.
Avoid release to the environment.
Wear eye protection, protective clothing, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Documents
Promotions

Promotions