Please login to your online account to display your discounted pricing

Silver Nitrate, Certified, 0.0450N ±0.0005N (0.045M), LabChem™

$335.14 - $335.14

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7732-18-5
CAS Max % 99.23
CAS 7761-88-8
CAS Max % 0.77
Purity Grade Certified Grade
Linear Formula AgNO3
Formula Weight 169.87
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC226604

 
lab chem
LC226604
4L Each for $335.14
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 169.87
Traceability to NIST Traceable to NIST
Linear Formula AgNO3
CAS Max % 99.23
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Quantity 4L
CAS 7732-18-5
Color Clear, colorless
Physical Form Liquid
CAS 7761-88-8
CAS Max % 0.77
Concentration 0.0450N ±0.0005N (0.045M)
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Density 1g/mL
Specific Gravity 1g/mL
Decomposition Information Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide
Packaging Amber Glass
Chemical Name or Material Silver Nitrate
Identification Passes Test
Storage Information Room Temperature
Solubility Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.