Please login to your online account to display your discounted pricing

Silver Nitrate ACS, Ricca Chemical


Quantity 100g
Packaging Poly wht WM

Manufacturer: ricca chemical company  RIRDCS0170100B1

 View more versions of this product

Catalog No. RDCS0170100

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Purity Grade ACS Grade
Physical Form Crystals
Color Transparent, colorless or white
CAS Min % 98.0
CAS Max % 100.0
CAS 7761-88-8
Packaging Poly wht WM
Chemical Name or Material Silver Nitrate
Shelf Life 60 months

Specifications

Chemical Name or Material Silver Nitrate
CAS 7761-88-8
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Grade
Physical Form Crystals
Accessories

60 months