Please login to your online account to display your discounted pricing

Silver Nitrate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7761-88-8
Purity Grade ACS Grade
Physical Form Crystalline
Molecular Formula AgNO3
Formula Weight 169.87
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK216916

 
avantor performance
216903
Glass 16OZ CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 169.87
Quantity 16OZ
Chemical Name or Material Silver Nitrate
Packaging Glass
Physical Form Crystalline
Molecular Formula AgNO3
CAS 7761-88-8