missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver nitrate, ACS, >99.9% (metals basis) , Alfa Aesar™

Silver nitrate, AgNO3, CAS Number-7761-88-8, silvernitrate, silver i nitrate, argenti nitras, nitric acid silver i salt, nitrate d'argent, lunar caustic, argerol, silver mononitrate, silver nitrate, silbernitrat, 100g, >99.9% (metals basis), CHEBI:32130, 4.352 g/mL

Manufacturer:  Thermo Fisher Scientific 1141422

 View more versions of this product

Catalog No. AA1141422



Description

Description

Silver nitrate, AgNO3, CAS Number-7761-88-8, silvernitrate, silver i nitrate, argenti nitras, nitric acid silver i salt, nitrate d'argent, lunar caustic, argerol, silver mononitrate, silver nitrate, silbernitrat, 100g, >99.9% (metals basis), CHEBI:32130, 4.352 g/mL

Specifications

Specifications

Silver nitrate
7761-88-8
AgNO3
argenti nitras, argerol, lunar caustic, nitrate d'argent, nitric acid silver i salt, silbernitrat, silver i nitrate, silver mononitrate, silver nitrate, silvernitrate
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
ACS Reagent
Light sensitive
100g
For AAS analysis
>99.9% (metals basis)
MFCD00003414
Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether.
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Crystalline
4.352 g/mL
212°C

Chemical Identifiers

7761-88-8
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver;nitrate
AgNO3
MFCD00003414
argenti nitras, argerol, lunar caustic, nitrate d'argent, nitric acid silver i salt, silbernitrat, silver i nitrate, silver mononitrate, silver nitrate, silvernitrate
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
Safety and Handling

Safety and Handling

P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H290-H314-H335-H373

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-853-9

RTECSNumber : VW4725000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents
Promotions

Promotions