missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.282N

Manufacturer:  Alfa Aesar™ 88615LQ

 View more versions of this product

Catalog No. AA88615LQ



Specifications

Specifications

Silver nitrate Acculute Standard Volumetric Solution
7761-88-8
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
Colorless
Light Sensitive
Final Concentration 0.282N
AgNO3
UN3093
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Liquid
1 unit

Chemical Identifiers

7761-88-8
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
AgNO3
MFCD00003414
24470
silver;nitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318-H290
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.

TSCA : TSCA

SDS Documents