missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver nitrate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Alfa Aesar™

Manufacturer:  Alfa Aesar 88614LS

 View more versions of this product

Catalog No. AA88614LS



Specifications

Specifications

Silver nitrate
Colourless liquid
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
Light sensitive
Colorless
7761-88-8
AgNO3
Silver nitrate: 40%; Water: 60%
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Liquid
23 hPa (17mm Hg) at 20°C
6 units

Chemical Identifiers

7761-88-8
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
AgNO3
MFCD00003414
24470
silver;nitrate
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H272-H314-H318-H290-H400-H410: May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-853-9

SDS Documents