missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver nitrate, 99.9995% (metals basis), Alfa Aesar™, Premion™

Manufacturer:  Thermo Fisher Scientific 1085806

 View more versions of this product

Catalog No. AA1085806



Specifications

Specifications

Silver nitrate
99.9995% (metals basis)
MFCD00003414
argenti nitras
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
Powder
Odorless
7761-88-8
AgNO3
White crystals
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
169.87
4.352g/cm3 at 20°C
5g

Chemical Identifiers

7761-88-8
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver;nitrate
AgNO3
MFCD00003414
argenti nitras
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
Safety and Handling

Safety and Handling

H272-H314-H290-H400-H410: May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 5.1, Packaging Group: I

EINECSNumber : 231-853-9

Documents