Learn More
Silver nitrate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035601M4
Description
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver nitrate | |
Colorless | |
20 L | |
MFCD00003414 | |
Miscible with water,alcohol and aqueous ammonia. | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
Silver nitrate: 1.7%; Water: 98.3% | |
Light sensitive | |
23 hPa (17mm Hg) at 20°C |
7761-88-8 | |
Liquid | |
AgNO3 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
Odorless | |
Colourless liquid |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : DOT Class: 8; Packaging Group: III
EINECSNumber : 231-853-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only