missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, 0.1N in Isopropyl Alcohol, Ricca Chemical

$273.27 - $850.52

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.872
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver;nitrate
SMILES [N+](=O)([O-])[O-].[Ag+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7033-32 View Documents Ricca Chemical Company
RI703332
1L Amber Plastic Each for $273.27
7033-1 View Documents Ricca Chemical Company
RI70331
4L Amber Plastic Each for $850.52
Specifications

Chemical Identifiers

7761-88-8
169.872
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[N+](=O)([O-])[O-].[Ag+]
AgNO3
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver;nitrate

Specifications

7761-88-8
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver;nitrate
24470
Amber Plastic
Silver Nitrate
AgNO3
0.100N (N/10) in Isopropyl Alcohol
[N+](=O)([O-])[O-].[Ag+]
169.872
CHEBI:32130
1L