Please login to your online account to display your discounted pricing

Silver Nitrate, 0.10N N/10) Volumetric Solution, Standard, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Silver Nitrate, 0.10 Normal (N/10)
CAS 7761-88-8
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MK61421

 
avantor performance
614260
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK61424

 
avantor performance
614201
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Silver Nitrate, 0.10 Normal (N/10)
CAS 7761-88-8
Color Clear colorless
Physical Form Liquid