Please login to your online account to display your discounted pricing

Silver Nitrate, 0.100 Normal (N/10), Ricca Chemical

$1,435.97 - $1,435.97

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7761-88-8
CAS Min % 1.64
CAS Max % 1.7
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

70-003-2CS

 
ricca chemical company
RIR700000012X1B
Black poly 12L Each for $1,435.97
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 12L
CAS Min % 96.0
CAS Max % 100.0
Shelf Life 18 Months
Chemical Name or Material Silver Nitrate
Packaging Black poly
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10)
CAS 7761-88-8
CAS Min % 1.64
CAS Max % 1.7
Safety and Handling

18 Months