Please login to your online account to display your discounted pricing

Silver Nitrate, 0.0855N, 1mL = 5mg NaCl, for Chloride Titrations, Ricca Chemical

$421.12 - $421.12

Chemical Identifiers

Chemical Name or Material Silver Nitrate
Name Note 0.0855N, 1mL = 5mg NaCl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6980-1

 
ricca chemical company
RI69801
Amber Plastic 4L Each for $421.12
Description & Specifications

Specifications

Quantity 4L
Equivalence 1mL = 5mg NaCl
Chemical Name or Material Silver Nitrate
Name Note 0.0855N, 1mL = 5mg NaCl
Concentration or Composition (by Analyte or Components) 0.0855N
Packaging Amber Plastic

For Chloride Titrations